missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Apolipoprotein C2 Polyclonal Antibody
Goat Polyclonal Antibody
Marke: Invitrogen™ PA122561
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Apolipoproteins are protein components of plasma lipoproteins. The apolipoprotein C gene family encodes four homologous proteins designated apoC-I to -IV, which specifically modulate the metabolism of triglyceride-rich lipoproteins. The human apoC-I gene maps to chromosome 19q13.2 and is expressed primarily in the liver where it is activated when monocytes differentiate into macrophages. The human apoC-II gene maps to chromosome 19q13.2 and encodes a 79 amino acid single chain protein that is a necessary cofactor for the activation of lipoprotein lipase, the enzyme that hydrolyzes triglycerides in plasma and transfers the fatty acids to tissues.
Spezifikation
| Apolipoprotein C2 | |
| Polyclonal | |
| Unconjugated | |
| APOC2 | |
| APC 2; APC2; APO C2; Apo CII; APOC 2; ApoC II; Apoc2; APOC2 protein; ApoCII; APO-CII; apoC-II; Apolipoprotein C II; apolipoprotein C2; Apolipoprotein CII; Apolipoprotein C-II; ApolipoproteinCII; MGC75082; ProapoC-II; Proapolipoprotein C-II; RGD1560725 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 344 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| BBS with 5mM EDTA, 0.075M NaCl and 0.01% sodium azide; pH 8 | |
| P02655 | |
| APOC2 | |
| Apolipoprotein Type C-II produced synthetically in full-length form (not selected epitopes). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction