Learn More
Invitrogen™ APC Monoclonal Antibody (eBioAPC-6A2), Biotin, eBioscience™

Mouse Monoclonal Antibody
172.00 CHF
Spezifikation
| Antigen | APC |
|---|---|
| Klon | eBioAPC-6A2 |
| Konzentration | 0.5 mg/mL |
| Inhalt und Lagerung | 4°C, store in dark, DO NOT FREEZE! |
| Anwendungen | Flow Cytometry |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15269799
![]() |
Invitrogen™
13-4170-82 |
100 μg |
172.00 CHF
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Description: The eBioAPC-6A2 monoclonal antibody reacts with allophycocyanin (APC) and APC-Cy7, with no crossreactivity to fluorescein (FITC) or phycoerythrin (PE). When using eBioAPC-6A2 as a second-step antibody, in combination with an appropriate third step, to bind a APC-conjugated primary antibody, there is no decrease in the fluorescent intensity of the directly conjugated APC antibody. Therefore no interference or quenching of the APC fluorochrome is observed. The eBioAPC-6A2 can be used for separation of cells labeled with APC-conjugated antibodies or for staining. Please note that the optimal concentration for this antibody will vary depending on the density of the primary APC-conjugated antibody and the application tested. Applications Reported: This eBioAPC-6A2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioAPC-6A2 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells stained with an APC-conjugated antibody. This can be used at less than or equal to 0.015 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
Anti-APC (Allophycocyanin) antibodies can be used to bind APC-conjugated primary antibodies without decreasing fluorescent intensity of the APC fluorochrome. This method is used for separation of cells labeled with APC-conjugated antibodies.Spezifikation
| APC | |
| 0.5 mg/mL | |
| Flow Cytometry | |
| Biotin | |
| Mouse | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Primary | |
| Affinity Chromatography |
| eBioAPC-6A2 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Tag | |
| APC | |
| Antibody |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.