Learn More
APC Anti-Human CD16 (3G8), Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech APC-65090-100TESTS
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The CD16 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD16. This antibody recognizes human antigen. The CD16 antibody has been validated for the following applications: FC analysis. This antibody is apc.Spezifikation
| CD16 | |
| 3G8 | |
| APC | |
| Mouse | |
| 100 Tests | |
| 2214 | |
| Store at 2-8°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry | |
| 5 μl/test | |
| CD16; CD16A; FCG3; FCGR3; FCGRIII; FCR-10; FCRIII; FCRIIIA; IGFR3 | |
| Affinity purification | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG1 κ |