missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AOAH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12911-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Removes the secondary (acyloxyacyl-linked) fatty acyl chains from the lipid A region of bacterial lipopolysaccharides.Spezifikation
AOAH | |
Polyclonal | |
Unconjugated | |
AOAH | |
Acyloxyacyl hydrolase, AOAH | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27052, 313, 498757 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35298, P28039 | |
AOAH | |
AOAH Fusion Protein Ag3892 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |