missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF202 Monoclonal Antibody (1E9), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA520291
Additional Details : Gewicht : 0.00010kg
Beschreibung
Peptide Sequence: QEPQETQEPE ILSFTYTGDR SKDEEECLEQ EDLSLEDIHR PVLGEPEIHQ TPDWEIVFED NPGRLNERRF GTNISQVNSF VNLRETTPVH PLLGRHHDCS
ZNF202 Monoclonal antibody specifically detects ZNF202 in Human samples. It is validated for ELISA, Immunohistochemistry (PFA fixed), Western Blot.Spezifikation
ZNF202 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
O95125 | |
ZNF202 | |
ZNF202 (NP_003446, 301 a.a. ∽ 400 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
1E9 | |
Unconjugated | |
ZNF202 | |
Zinc finger protein 202; zinc finger protein with KRAB and SCAN domains 10; ZKSCAN10; ZNF202; ZSCAN42 | |
Mouse | |
Affinity chromatography | |
RUO | |
7753 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |