missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-ZFYVE16, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Marke: Abnova H00009765-AP22.1Set
Includes: Capture antibody: rabbit MaxPab affinity purified polyclonal anti-ZFYVE16 (100μg) , Detection antibody: mouse purified polyclonal anti-ZFYVE16 (20μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Spezifikation
| ZFYVE16 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ZFYVE16. | |
| DKFZp686E13162/ENDOFIN/KIAA0305 | |
| Mouse, Rabbit | |
| RUO | |
| 9765 | |
| Human | |
| Liquid |
| ELISA | |
| Unconjugated | |
| ZFYVE16 | |
| ZFYVE16 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-ZFYVE16 (100 μg) 2. Detection antibody: mouse purified polyclonal anti-ZFYVE16 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur