Learn More
vanin 3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant VNN3.
Marke: Abnova H00055350-A01.50uL
Description
This gene is the central gene in a cluster of three vanin genes on chromosome 6q23-q24. The open reading frame is disrupted by a frameshift, and all splice variants that have been described are candidates for nonsense-mediated decay (NMD). Consequently, it is unlikely that this gene expresses a protein in vivo, so it is classified as a pseudogene. Extensive alternative splicing has been described; the two most common variants are represented as RefSeqs. [provided by RefSeq
Sequence: ARYHKYNLFAPEIQFDFPKDSELVTFDTPFGKFGIFTCFDIFSHDPAVVVVDEFQLTAFSTPQHGTTRCPSSRLFPSIQHGPRPWESIYLLQIPTTPACTSpecifications
| vanin 3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant VNN3. | |
| VNN3 | |
| HSA238982/MGC171203/PAGEL-beta/PAGEL-eta/PAGEL-zeta | |
| Mouse | |
| 50 μL | |
| Yes | |
| 55350 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_018399 | |
| VNN3 | |
| VNN3 (NP_060869.2, 175 a.a. ∼ 274 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.