Learn More
Vδ 4 TCR Hamster anti-Mouse, FITC, Clone: GL2, BD
Hamster Monoclonal Antibody
Marke: BD Biosciences 552143
Additional Details : Gewicht : 0.01000kg
Beschreibung
The GL2 antibody reacts with Vδ 4 T-cell Receptor (TCR)-bearing T cells, which are the predominant γδ TCR-bearing cells in the adult intestinal epithelium (γδ IEL). The frequency of Vδ 4 TCR-bearing γδ IEL differs among inbred strains of mice and may be influenced by haplotypes of the MHC, Tcrg, and/or Tcrd gene complexes. There is evidence that these γδ IEL develop in the absence of thymic influence. T lymphocytes expressing Vδ 4 TCR have also been found in lactating mammary glands, the spleen, and the thymus.
Host Species: Hamster
Clone: GL2
Isotype: IgG2 κ
Species Reactivity: Mouse
Immunogen: G57BL/6 mouse intestinal intra-epithelial lymphocytes
Flow Cytometry
Spezifikation
Vδ 4 TCR | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
G57BL/6 mouse intestinal intra-epithelial lymphocytes | |
0.1 mg | |
Immunology | |
Mouse | |
IgG2 κ |
Flow Cytometry | |
GL2 | |
FITC | |
Hamster | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.