Learn More
ubiquitin-conjugating enzyme E2D 2 (UBC4/5 homolog, yeast), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant UBE2D2.
Marke: Abnova H00007322-A01.50uL
Beschreibung
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme functions in the ubiquitination of the tumor-suppressor protein p53, which is induced by an E3 ubiquitin-protein ligase. Two alternatively spliced transcript variants have been found for this gene and they encode distinct isoforms. [provided by RefSeq
Sequence: MALKRIHKELNDLARDPPAQCSAGPVGDDMFHWQATIMGPNDSPYQGGVFFLTIHFPTDYPFKPPKVAFTTRIYHPNINSNGSICLDILRSQWSSpezifikation
| ubiquitin-conjugating enzyme E2D 2 (UBC4/5 homolog, yeast) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant UBE2D2. | |
| UBE2D2 | |
| E2(17)KB2/PUBC1/UBC4/UBC4/5/UBCH5B | |
| Mouse | |
| 50 μL | |
| Ubiquitin/Proteasome | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_003339 | |
| UBE2D2 | |
| UBE2D2 (NP_003330, 1 a.a. ∼ 94 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 7322 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.