missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyrosine Hydroxylase Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518372
Additional Details : Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with canine and rat based on sequence homology.
Tyrosine Hydroxylase Polyclonal antibody specifically detects Tyrosine Hydroxylase in Human samples. It is validated for Immunohistochemistry.Spezifikation
Tyrosine Hydroxylase | |
Polyclonal | |
Unconjugated | |
TH | |
dystonia 14; DYT14; DYT5b; EC 1.14.16.2; HGNC:11782; Th; TH isoform 3; TH isoform a; th1; TH-4; The; TY3H; TYH; TYH antibody; Tyrosine 3-hydroxylase; tyrosine 3-monooxygenase; tyrosine hydroxylase | |
Goat | |
Antigen affinity chromatography | |
RUO | |
7054 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P07101 | |
TH | |
Synthetic peptide sequence (VQDELDTLAHAL) corresponding to the C-terminus amino acids of TH | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |