Learn More
anti-TRIM55 Matched Antibody Pair, Abnova
Marke: Abnova H00084675-AP21.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-TRIM55 (100μg); Mouse purified polyclonal anti-TRIM55 (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Beschreibung
The protein encoded by this gene contains a RING zinc finger, a motif known to be involved in protein-protein interactions. This protein associates transiently with microtubules, myosin, and titin during muscle sarcomere assembly. It may act as a transient adaptor and plays a regulatory role in the assembly of sarcomeres. Four alternatively spliced transcript variants encoding distinct isoforms have been described.
ELISA
Spezifikation
| TRIM55 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TRIM55. | |
| MURF-2/RNF29 | |
| Mouse, Rabbit | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-TRIM55 (100 μg) 2. Detection antibody: mouse purified polyclonal anti-TRIM55 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Unconjugated | |
| TRIM55 | |
| TRIM55 | |
| Affinity Purified | |
| RUO | |
| 84675 | |
| Human |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.