Learn More
TREH Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA522076
Additional Details : Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, mouse stomach. Predicted reactivity: Mouse (80%), Rat (80%), Rabbit (82%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TREH Polyclonal antibody specifically detects TREH in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
TREH | |
Polyclonal | |
Unconjugated | |
TREH | |
2210412M19Rik; Alpha,alpha-trehalase; alpha,alpha-trehalose glucohydrolase; LOW QUALITY PROTEIN: trehalase; trehalase; si:ch211-147p17.2; TRE; TREA; TREH; Trehalase; trehalase (brush-border membrane glycoprotein) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11181, 58866 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O43280, Q9JLT2 | |
TREH | |
Recombinant fragment corresponding to a region within amino acids 7 and 285 of Trehalase (Uniprot ID#O43280) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |