Learn More
TRA-1-81 Antigen Mouse anti-Human, PE, Clone: TRA-1-81, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 560885
Additional Details : Gewicht : 0.01000kg
Beschreibung
The TRA-1-81 monoclonal antibody reacts with a pluripotent-stem-cell-specific epitope on a high-molecular-weight transmembrane glycoprotein. The TRA-1-81 antigen is an epitope on the same keratan sulfate core molecule, podocalyxin, as 4 other distinct antigens on tumor-derived cell lines, TRA-1-60, GCTM2, K4, and K21. The expression of TRA-1-81 antigen is stage-specific and can be used to characterize embryonic cells and monitor their differentiation. The antigen is found on teratocarcinoma (embryonal carcinoma or EC), embryonic inner cell mass (but not morula or trophoblast), and embryonic stem (ES) cells. As human EC and ES cells undergo differentiation, expression of TRA-1-81 antigen is lost.
Spezifikation
TRA-1-81 Antigen | |
Monoclonal | |
PE | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
TRA-1-81 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human Embryonal Carcinoma Cell Line | |
25 Tests | |
Cell Biology | |
Human | |
IgM κ |
For Research Use Only.