missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-TPP1 Matched Antibody Pair, Abnova™
Marke: Abnova H00001200-AP51.1Set
Additional Details : Gewicht : 0.00010kg
Includes: Capture antibody: mouse monoclonal anti-TPP1 (100μg); Detection antibody: rabbit purified polyclonal anti-TPP1 (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Beschreibung
Host Species: Mouse, Rabbit
Cross Reactivity: Human
ELISA Pair
Spezifikation
TPP1 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TPP1. | |
CLN2/GIG1/LPIC/MGC21297 | |
Mouse, Rabbit | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-TPP1 (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-TPP1 (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
TPP1 | |
TPP1 | |
1 Set | |
1200 | |
Human |