Learn More
TNFRSF17, Human, Matched Antibody Pair (AP41), Abnova™
Artikelnummer. 16171774
Change view
Click to view available options
Menge:
1 Set
Packungsgröße:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16171774 1 Set 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 16171774 Lieferant Abnova Lieferanten-Nr. H00000608AP41.1Set
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Includes
Capture antibody: mouse monoclonal anti-TNFRSF17 (100μg) , Detection antibody: biotinylated mouse monoclonal anti-TNFRSF17, IgG1 Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
To detect and quantify protein level of human
Spezifikation
Antigen TNFRSF17
Anwendungen ELISA
Klassifikation Monoclonal/Polyclonal
Konjugat Biotin
Beschreibung This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TNFRSF17.
Gen TNFRSF17
Gen-Alias BCM/BCMA/CD269
Gensymbole TNFRSF17
Wirtsspezies Mouse
Menge 1 Set
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 608
Enthält Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-TNFRSF17 (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-TNFRSF17, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Zielspezies Human
Inhalt und Lagerung Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid