missing translation for 'onlineSavingsMsg'
Learn More

TNF Rat anti-Mouse, APC, Clone: MP6-XT22, BD

Rat Monoclonal Antibody

Marke:  BD Biosciences 554420

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15827688

  • 369.00 CHF / 1 Stück


Verfügbar ab
03-06-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The MP6-XT22 antibody reacts with mouse tumor necrosis factor (TNF, also known as TNF-α). The immunogen used to generate this hybridoma was recombinant mouse TNF.

Host Species: Rat
Clone: MP6-XT22
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Recombinant mouse TNF

Intracellular Staining

Spezifikation

Spezifikation

TNF
MP6-XT22
APC
Affinity Purified
Recombinant mouse TNF
0.1mg
Immunology
Primary
Murine
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG1
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

For Research Use Only.