Learn More
TNF Mouse anti-Human, APC, Clone: MAB11, BD
Mouse Monoclonal Antibody
605.00 CHF - 660.00 CHF
Spezifikation
Antigen | TNF |
---|---|
Klon | MAB11 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | APC |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15857748
|
BD Biosciences
554514 |
0.1 mg |
660.00 CHF
1 Stück |
Verfügbar ab: 07-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15886628
|
BD Biosciences
551384 |
50 Tests |
605.00 CHF
1 Stück |
Verfügbar ab: 13-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The MAb11 antibody reacts with human tumor necrosis factor (TNF, formerly known as TNF-α) protein. TNF is an efficient paracrine and endocrine mediator of inflammatory and immune functions. It regulates the growth and differentiation of a variety of cell types. TNF is cytotoxic for transformed cells when in conjunction with IFN-γ. It is secreted by activated monocytes/macrophages and other cells such as B cells, T cells and fibroblasts. The immunogen used to generate the MAb11 hybridoma was recombinant human TNF. The use of the MAb11 antibody has been reported to cross-react with TNF of rhesus monkey.
Host Species: Mouse
Clone: MAb11
Isotype: IgG1 κ
Species Reactivity: Non-human primate
Immunogen: Recombinant Human TNF
Intracellular Staining
Spezifikation
TNF | |
Flow Cytometry | |
APC | |
RUO | |
Recombinant Human TNF | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
MAB11 | |
Monoclonal | |
Mouse | |
Immunology | |
IgG1 κ | |
Affinity Purified |
For Research Use Only.