Learn More
TRIF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528105
Additional Details : Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa. Predicted reactivity: Rhesus Monkey (100%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TRIF Polyclonal antibody specifically detects TRIF in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
TRIF | |
Polyclonal | |
Unconjugated | |
TICAM1 | |
AW046014; AW547018; hCG_39321; HSD34; IIAE6; MyD88-3; Proline-rich, vinculin and TIR domain-containing protein B; PRVTIRB; Putative NF-kappa-B-activating protein 502H; RNF36; TI; Ticam1; TICAM-1; TIR domain containing adaptor inducing interferon-beta; TIR domain-containing adapter molecule 1; TIR domain-containing adapter protein inducing IFN-beta; TIR domain-containing adaptor inducing interferon-beta; TIR-domain containing adaptor inducing interferon-beta; toll like receptor adaptor molecule 1; toll-interleukin 1 receptor domain-containing adapter protein inducing interferon-beta; toll-interleukin I receptor domain containing adaptor molecule 1; toll-interleukin-1 receptor domain-containing adapter protein inducing interferon beta; toll-like receptor adaptor molecule 1; TRIF | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
148022 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1.01 mg/mL | |
PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q8IUC6 | |
TICAM1 | |
Synthetic peptide corresponding to a region within amino acids 650 and 712 of Human TICAM1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |