Learn More
anti-TGIF2, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Marke: Abnova H00060436-AP41.1Set
Includes: Capture antibody: mouse monoclonal anti-TGIF2, IgG2a Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-TGIF2, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Spezifikation
| TGIF2 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TGIF2. | |
| TGIF2 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-TGIF2, IgG2a Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-TGIF2, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Biotin | |
| TGIF2 | |
| Mouse | |
| RUO | |
| 60436 | |
| Human | |
| Liquid |