Learn More
anti-TDP1 Matched Antibody Pair, Abnova
Artikelnummer. 16065249
Change view
Click to view available options
Menge:
1 Set
Packungsgröße:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16065249 1 Set 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 16065249 Lieferant Abnova Lieferanten-Nr. H00055775AP11.1Set
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Includes
Rabbit MaxPab™ affinity purified polyclonal anti-TDP1 (100μg); Mouse monoclonal anti-TDP1, IgG1κ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
The protein encoded by this gene is involved in repairing stalled topoisomerase I-DNA complexes by catalyzing the hydrolysis of the phosphodiester bond between the tyrosine residue of topoisomerase I and the 3-prime phosphate of DNA. This protein may also remove glycolate from single-stranded DNA containing 3-prime phosphoglycolate, suggesting a role in repair of free-radical mediated DNA double-strand breaks. This gene is a member of the phospholipase D family and contains two PLD phosphodiesterase domains. Mutations in this gene are associated with the disease spinocerebellar ataxia with axonal neuropathy (SCAN1). While several transcript variants may exist for this gene, the full-length natures of only two have been described to date. These two represent the major variants of this gene and encode the same isoform.
ELISA
Spezifikation
Antigen TDP1
Anwendungen ELISA
Klassifikation Monoclonal/Polyclonal
Konjugat Unconjugated
Beschreibung This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TDP1.
Gen TDP1
Gen-Alias FLJ11090/MGC104252
Gensymbole TDP1
Wirtsspezies Mouse, Rabbit
Reinigungsverfahren Affinity Purified
Menge 1 Set
Regulatorischer Status RUO
Forschungsgebiet Stem Cell Biology
Primär oder sekundär Primary
Gen-ID (Entrez) 55775
Enthält Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-TDP1 (100 μg) 2. Detection antibody: mouse monoclonal anti-TDP1, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Zielspezies Human
Inhalt und Lagerung Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Isotype IgG1