Learn More
TCFL5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA124394
Additional Details : Gewicht : 0.01000kg
Beschreibung
PA1-24394 detects TCFL5 in Human samples. PA1-24394 has been successfully used in Western Blot procedures. PA1-24394 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human TCFL5. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
TCFL5 is a new bHLH transcription factor that negatively regulates upstream transcription factor-dependent transcription.Spezifikation
TCFL5 | |
Polyclonal | |
Unconjugated | |
Q9UL49 | |
TCFL5 | |
Synthetic peptide region of human TCFL5. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TCFL5 | |
CHA, E2BP-1, Figlb, MGC46135 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10732 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |