missing translation for 'onlineSavingsMsg'
Learn More

TCEAL2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA543626

Additional Details : Gewicht : 0.00010kg

Artikelnummer. 15596142

  • 604.00 CHF / 100 Mikroliter
Verfügbar ab: 19-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Peptide sequence: MEKLFNENEG MPSNQGKIDN EEQPPHEGKP EVACILEDKK LENEGNTENT Sequence homology: Human: 100%

TCEAL2 Polyclonal antibody specifically detects TCEAL2 in Human samples. It is validated for Western Blot.
Spezifikation

Spezifikation

TCEAL2
Polyclonal
Unconjugated
TCEAL2
my048; MY0876G0; MY0876G05; TCEAL2; TCEA-like protein 2; transcription elongation factor A (SII)-like 2; transcription elongation factor A like 2; transcription elongation factor A protein-like 2; Transcription elongation factor S-II protein-like 2; WEX1
Rabbit
Affinity Chromatography
RUO
140597
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9H3H9
TCEAL2
Synthetic peptide directed towards the N-terminal of human TCEAL2
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote