missing translation for 'onlineSavingsMsg'
Learn More

TC10 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA525749

Additional Details : Gewicht : 0.01000kg

Artikelnummer. 13259388

  • 658.00 CHF / 100 Mikrogramm
Verfügbar ab: 11-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This antibody is predicted to react with bovine, chicken, canine, equine, mouse, porcine, non-human primate, rat and yeast based on sequence homology.

TC10 Polyclonal antibody specifically detects TC10 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

TC10
Polyclonal
Unconjugated
RHOQ
ARHQ; epididymis secretory protein Li 42; HEL-S-42; ras homolog family member Q; ras homolog gene family, member Q; RASL7A; ras-like protein family member 7A; Ras-like protein TC10; RAS-like, family 7, member A; ras-related GTP-binding protein TC10; RHOQ; Rho-related GTP-binding protein RhoQ; small GTPase Tc10; Tc10; TC10A
Rabbit
Antigen affinity chromatography, Protein A
RUO
23433
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
P17081
RHOQ
KLH conjugated synthetic peptide between 48-76 amino acids from the central region of human TC10
400 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote