missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS2R50 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen OSR00177W
Additional Details : Gewicht : 0.01000kg
Beschreibung
Reconstitute with 100 μL of distilled water. Centrifuge to remove any insoluble material.
TAS2R50 belongs to the large TAS2R receptor family. TAS2Rs are expressed on the surface of taste receptor cells and mediate the perception of bitterness through a G protein-coupled second messenger pathway. Receptor that may play a role in the perception of bitterness and is gustducin-linked. May play a role in sensing the chemical composition of the gastrointestinal content. The activity of this receptor may stimulate alpha gustducin, mediate PLC-beta-2 activation and lead to the gating of TRPM5. Expressed in subsets of taste receptor cells of the tongue and exclusively in gustducin-positive cells. Most taste cells may be activated by a limited number of bitter compounds; individual taste cells can discriminate among bitter stimuli.Spezifikation
TAS2R50 | |
Polyclonal | |
TAS2R50 | |
Taste receptor type 2 member 50, T2R50, T2R51, TAS2R50, MGC138305 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
P59544 | |
TAS2R50 | |
Synthetic peptide from human taste receptor, type 2, member 50. | |
RUO | |
259296 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |