missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS2R3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA539712
Additional Details : Gewicht : 0.00900kg
Beschreibung
This gene encodes a member of a family of candidate taste receptors that are members of the G protein-coupled receptor superfamily and that are specifically expressed by taste receptor cells of the tongue and palate epithelia. These apparently intronless taste receptor genes encode a 7-transmembrane receptor protein, functioning as a bitter taste receptor. This gene is clustered with another 3 candidate taste receptor genes in chromosome 7 and is genetically linked to loci that influence bitter perception. Gustducin-coupled receptor implicated in the perception of bitter compounds in the oral cavity and the gastrointestinal tract. Signals through PLCB2 and the calcium-regulated cation channel TRPM5. Expressed in subsets of taste receptor cells of the tongue and palate epithelium and exclusively in gustducin-positive cells. Expressed in the antrum and fundus (part of the stomach), duodenum and in gastric endocrine cells. Several bitter taste receptors are expressed in a single taste receptor cell.Spezifikation
TAS2R3 | |
Polyclonal | |
Unconjugated | |
TAS2R3 | |
Taste receptor type 2 member 3, T2R3, TAS2R3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
50831 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9NYW6 | |
TAS2R3 | |
A synthetic peptide derived from the internal region of human TAS2R3 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |