missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tankyrase 2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA517930
Additional Details : Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with canine, mouse and rat based on sequence homology.
Tankyrase 2 Polyclonal antibody specifically detects Tankyrase 2 in Human samples. It is validated for Western Blot.Spezifikation
Tankyrase 2 | |
Polyclonal | |
Unconjugated | |
TNKS2 | |
5430432P15Rik; AA517131; ADP-ribosyltransferase diphtheria toxin-like 6; AI662480; ARTD6; PARP5B; PARP-5b; PARP5C; PARP-5c; pART6; Poly [ADP-ribose] polymerase 5B; Poly [ADP-ribose] polymerase tankyrase-2; Protein poly-ADP-ribosyltransferase tankyrase-2; TANK2; tankyrase 2; tankyrase II; tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase 2; tankyrase-2; Tankyrase-like protein; tankyrase-related protein; TNKL; tnks 2; TNKS2; TNKS-2; TRF1-interacting ankyrin-related ADP-ribose polymerase 2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
80351 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9H2K2 | |
TNKS2 | |
Synthetic peptide sequence (RECOMBINANT2) corresponding to the internal amino acids of TNKS2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |