missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Talin 1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA519186
Additional Details : Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology.
Talin 1 Polyclonal antibody specifically detects Talin 1 in Human samples. It is validated for Western Blot.Spezifikation
Talin 1 | |
Polyclonal | |
Unconjugated | |
TLN1 | |
ILWEQ; KIAA1027; talin 1; talin-1; TLN; TLN1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
7094 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9Y490 | |
TLN1 | |
Synthetic peptide sequence (QEALHTQMLTAVQE) corresponding to the internal amino acids of TLN1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |