missing translation for 'onlineSavingsMsg'
Learn More

TAF5L Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA536309

Additional Details : Gewicht : 0.00910kg

Artikelnummer. 15587611

  • 602.00 CHF / 100 Mikroliter
Verfügbar ab: 30-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This antibody detects endogenous protein at a molecular weight of 66 kDa. Purity is >95% by SDS-PAGE.

TAF5L Polyclonal antibody specifically detects TAF5L in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin).
Spezifikation

Spezifikation

TAF5L
Polyclonal
Unconjugated
Taf5l
1110005N04Rik; AI849020; Paf65b; PAF65-beta; PCAF associated factor 65 beta; PCAF-associated factor 65 beta; TAF5L; TAF5-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 5L; TAF5-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor; TAF5-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor, 65kDa; TATA box binding protein associated factor 5 like; TATA-box binding protein associated factor 5 like
Rabbit
Antigen affinity chromatography
RUO
102162, 27097, 307927
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
O75529, Q91WQ5
Taf5l
Synthetic peptide corresponding to amino acids 301-350 of human TAF5L
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote