Learn More
T- and B-Cell Activation Antigen Rat anti-Mouse, FITC, Clone: GL7, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 562080
Additional Details : Gewicht : 0.01000kg
Beschreibung
The GL7 antibody reacts with a 35-kDa cell-surface protein found on T and B lymphocytes activated in vitro,on bone marrow Pre-B-II cells, germinal-center B cells, and a subpopulation of the thymocyte fraction expressing high density of CD3e. There is strain variability with respect to antigen distribution on thymocytes and Con A-activated spleen cells, with expression in BALB/c greater than that in C57BL/6.
Spezifikation
T- and B-Cell Activation Antigen | |
Monoclonal | |
0.5mg/mL | |
GL7 | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgM κ |
Flow Cytometry | |
GL7 | |
FITC | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
In vitro-activated T-cell-depleted CBA/Ca mouse splenocytes | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.