Learn More
anti-SUOX Matched Antibody Pair, Abnova
Marke:  Abnova H00006821-AP21.1Set
Artikelnummer. 16132694
  • ONLINE EXCLUSIVE PRICE
    901.00 CHF
    783.30 CHF / 1 Set
    Sparen 117.70 CHF
    13% Rabatt
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-SUOX (100μg); Mouse purified polyclonal anti-SUOX (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Beschreibung
Beschreibung
Sulfite oxidase is a homodimeric protein localized to the intermembrane space of mitochondria. Each subunit contains a heme domain and a molybdopterin-binding domain. The enzyme catalyzes the oxidation of sulfite to sulfate, the final reaction in the oxidative degradation of the sulfur amino acids cysteine and methionine. Sulfite oxidase deficiency results in neurological abnormalities which are often fatal at an early age. Alternative splicing results in multiple transcript variants encoding identical proteins.
ELISA
Spezifikation
Spezifikation
SUOX
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human SUOX.
SUOX
Affinity Purified
RUO
6821
Human
ELISA
Unconjugated
SUOX
Mouse, Rabbit
1 Set
Primary
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-SUOX (100 μg) 2. Detection antibody: mouse purified polyclonal anti-SUOX (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.