missing translation for 'onlineSavingsMsg'
Learn More

Stat2 Mouse anti-Human, Unlabeled, Clone: 22, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 610187

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15825319

  • 280.00 CHF / 1 Set


Verfügbar ab
01-06-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The Stat proteins function as both cytoplasmic signal transducers and activators of transcription. Stat91/84 (the two proteins are the result of alternate splicing-Stat91 has an additional 38 C-terminal amino acids) and Stat113 were the first identified members of this protein family. These three polypeptides contain both SH2 and SH3 domains and have also been described as members of the ISGF3 (interferon-stimulated gene factor 3) complex. With the discovery of additional members of the Stat family (Stats3 & 4), the nomenclature has been revised to indicate the Stat family members in the order of their discovery. Stat 91, 84, and 113 have become Stat1α, Stat1β, and Stat2, respectively. In response to IFN-α treatment, Stat1α, Stat1β, and Stat2 become tyrosine-phosphorylated and migrate to the nucleus where they join a 48kDa DNA binding protein and subsequently direct the transcription at IFN-α responsive elements. The genes encoding Stat2 and Stat1 are closely related. An extended acidic C-terminal region exclusive to Stat2 implicates a potential role in gene activation. Stat2 phosphorylation can occur independently of Stat1α, but phosphorylated Stat2 does appear to be dependent on its interaction with Stat1α for efficient nuclear localization and/or stability.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Spezifikation

Spezifikation

Stat2
22
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

For Research Use Only.