Learn More
SRC3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA1845
Additional Details : Gewicht : 0.01000kg
Beschreibung
PA1-845 detects amplified in breast cancer 1 (AIB1) from human tissues and cells. PA1-845 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ∽155 kDa protein representing AIB1 from T47D cell lysate. The PA1-845 immunogen is a synthetic peptide corresponding to residues G(3) L G E N L D P L A S D S(15) C of human AIB1. PA1-845 can be used with blocking peptide PEP-049.
SRC3 Polyclonal antibody specifically detects SRC3 in Human samples. It is validated for Western Blot.Spezifikation
SRC3 | |
Polyclonal | |
Unconjugated | |
NCOA3 | |
2010305B15Rik; Actr; AIB1; AIB-1; amplified in breast cancer 1 protein; amplified in breast cancer-1 protein homolog; AW321064; BHLHE42; CAGH16; CBP-interacting protein; class E basic helix-loop-helix protein 42; CTG26; KAT13B; MGC141848; NCOA3; NCoA-3; Nuclear receptor coactivator 3; OTTHUMP00000031716; p/Cip; pCip; Rac3; RAC-3; Receptor-associated coactivator 3; RP5-1049G16.3; Src3; SRC-3; steroid receptor coactivator protein 3; Thyroid hormone receptor activator molecule 1; TNRC14; TNRC16; Tram1; Tram-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8202 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
Q9Y6Q9 | |
NCOA3 | |
Synthetic peptide corresponding to residues G(3) L G E N L D P L A S D S(15) C of human AIB1. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |