Learn More
SOX13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA521556
Additional Details : Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (94%), Rat (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SOX13 Polyclonal antibody specifically detects SOX13 in Human samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Spezifikation
SOX13 | |
Polyclonal | |
Unconjugated | |
SOX13 | |
AA407916; AW259412; AW540933; ICA12; islet cell antibody 12; islet cell antigen 12; MGC117216; mSox13; RP11-74C13.1; SOX13; Sox-13; SRY (Sex determining region Y)-box 13; SRY box 13; SRY-box 13; SRY-box containing gene 13; SRY-related HMG-box gene 13; transcription factor SOX-13; Type 1 diabetes autoantigen ICA12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9580 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9UN79 | |
SOX13 | |
Recombinant fragment corresponding to a region within amino acids 75 and 520 of SOX13 (Uniprot ID#Q9UN79) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |