missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SORL1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA517941
Additional Details : Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, canine, human and rat based on sequence homology.
SORL1 Polyclonal antibody specifically detects SORL1 in Human samples. It is validated for Immunocytochemistry.Spezifikation
SORL1 | |
Polyclonal | |
Unconjugated | |
SORL1 | |
2900010L19Rik; AI596264; AW261561; C11orf32; FLJ21930; FLJ39258; gp250; LDLR relative with 11 ligand-binding repeats; low-density lipoprotein receptor relative with 11 ligand-binding repeats; Lr11; LRP9; mosaic protein LR11; mSorLA; RGD619914; Sorl1; SorLA; SorLA-1; sortilin related receptor 1; sortilin-related receptor; sortilin-related receptor, L(DLR class) A repeats containing; sortilin-related receptor, LDLR class A repeats-containing; sorting protein-related receptor containing LDLR class A repeats | |
Goat | |
Antigen affinity chromatography | |
RUO | |
6653 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q92673 | |
SORL1 | |
Synthetic peptide sequence (TGFSDDVPMVIA) corresponding to the C-terminus amino acids of SORL1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |