Learn More
anti-SNUPN Matched Antibody Pair, Abnova
Marke: Abnova H00010073-AP21.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-SNUPN (100μg); Mouse polyclonal anti-SNUPN (40μL); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Beschreibung
The nuclear import of the spliceosomal snRNPs U1, U2, U4 and U5, is dependent on the presence of a complex nuclear localization signal. The latter is composed of the 5′-2,2,7-terminal trimethylguanosine (m3G) cap structure of the U snRNA and the Sm core domain. The protein encoded by this gene interacts specifically with m3G-cap and functions as an snRNP-specific nuclear import receptor. Alternatively spliced transcript variants encoding the same protein have been identified for this gene.
ELISA
Spezifikation
| SNUPN | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human SNUPN. | |
| KPNBL/RNUT1/Snurportin1 | |
| Mouse, Rabbit | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-SNUPN (100 μg) 2. Detection antibody: mouse polyclonal anti-SNUPN (40 ul) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Unconjugated | |
| SNUPN | |
| SNUPN | |
| Affinity Purified | |
| RUO | |
| 10073 | |
| Human | |
| Serum |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.