Learn More
Serine Racemase Mouse, Unlabeled, Clone: 29, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 612053
Additional Details : Gewicht : 0.01000kg
Beschreibung
The majority of synapses in the central nervous system utilize glutamate as a neurotransmitter to produce rapid neuronal excitation. Glutamate has a diverse array of receptors that can be categorized into two groups: ionotropic and metabotropic. The ionotropic receptors are subdivided into two distinct types: 1) receptors for N-methyl D-aspartate (NMDAR) and 2) non-NMDA receptors for AMPA and kainate. NMDA receptors require coactivation at both glutamate and glycine sites. D-serine is a D-amino acid found in mammalian tissues that can act as a potent ligand for the glycine site on NMDA receptors. D-serine is made by the enzyme serine racemase, which is a member of the pyridoxal-5′ phosphate (PLP)-dependent enzyme family. Serine racemase mRNA is expressed in brain and liver, and serine racemase protein is expressed in glial cells. Degradation of D-serine by D-amino acid oxidase attenuates NMDA receptor-mediated calcium influx, and implicates D-serine as an endogenouse modulator of NMDA receptor function. Thus, glial cell production of D-serine via serine racemase activity may be an important system for modulation of synaptic transmission.
Host Species: Mouse
Clone: 29
Isotype: IgG1
Species Reactivity [for Features Main]: Mouse
Immunogen: Mouse serine racemase aa. 127-248
Immunofluorescence, Western Blotting
Spezifikation
Serine Racemase | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Mouse serine racemase aa. 127-248 | |
150 μg | |
Cell Biology | |
Mouse, Rat | |
IgG1 |
Western Blot | |
29 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.