Learn More
anti-SELE Matched Antibody Pair, Abnova
Marke: Abnova H00006401-AP21.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-SELE (100μg); Mouse purified polyclonal anti-SELE (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Beschreibung
The protein encoded by this gene is found in cytokine-stimulated endothelial cells and is thought to be responsible for the accumulation of blood leukocytes at sites of inflammation by mediating the adhesion of cells to the vascular lining. It exhibits structural features such as the presence of lectin- and EGF-like domains followed by short consensus repeat (SCR) domains that contain 6 conserved cysteine residues. These proteins are part of the selectin family of cell adhesion molecules. Adhesion molecules participate in the interaction between leukocytes and the endothelium and appear to be involved in the pathogenesis of atherosclerosis.
ELISA
Spezifikation
| SELE | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human SELE. | |
| CD62E/ELAM/ELAM1/ESEL/LECAM2 | |
| Mouse, Rabbit | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-SELE (100 μg) 2. Detection antibody: mouse purified polyclonal anti-SELE (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Unconjugated | |
| SELE | |
| SELE | |
| Affinity Purified | |
| RUO | |
| 6401 | |
| Human |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.