missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-S100A2 Matched Antibody Pair, Abnova™
Marke: Abnova H00006273-AP11.1Set
Additional Details : Gewicht : 0.00010kg
Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-S100A2 (100μg); Detection antibody: mouse monoclonal anti-S100A2, IgG2a Kappa (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Beschreibung
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human S100A2.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
ELISA Pair (Recombinant Protein)
Spezifikation
S100A2 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human S100A2. | |
CAN19/MGC111539/S100L | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-S100A2 (100 μg) 2. Detection antibody: mouse monoclonal anti-S100A2, IgG2a Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
S100A2 | |
S100A2 | |
Affinity Purified | |
RUO | |
6273 | |
Human |