Learn More
anti-RCVRN Matched Antibody Pair, Abnova
Marke: Abnova H00005957-AP21.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-RCVRN (100μg); Mouse polyclonal anti-RCVRN (40μL); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Beschreibung
This gene encodes a member of the recoverin family of neuronal calcium sensors. The encoded protein contains three calcium-binding EF-hand domains and may prolong the termination of the phototransduction cascade in the retina by blocking the phosphorylation of photo-activated rhodopsin. Recoverin may be the antigen responsible for cancer-associated retinopathy.
ELISA
Spezifikation
| RCVRN | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human RCVRN. | |
| RCV1 | |
| Mouse, Rabbit | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-RCVRN (100 μg) 2. Detection antibody: mouse polyclonal anti-RCVRN (40 ul) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Unconjugated | |
| RCVRN | |
| RCVRN | |
| Affinity Purified | |
| RUO | |
| 5957 | |
| Human | |
| Serum |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.