Learn More
F(ab')2-Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, PE, Invitrogen™
F(ab')2-Goat Polyclonal Secondary Antibody
Marke: Invitrogen A10544
Additional Details : Gewicht : 0.00450kg
Beschreibung
Rat IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Rat IgG (H+L) Cross-Adsorbed in Rat samples. It is validated for Flow Cytometry.Spezifikation
Rat IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
PE | |
Goat | |
purified | |
RUO | |
Rat | |
Liquid |
Flow Cytometry | |
1 mg/mL | |
PBS with 5mM sodium azide; pH 7.5 | |
Gamma Immunoglobins Heavy and Light chains | |
250 μL | |
Secondary | |
4° C | |
IgG |