missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PYHIN1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA525293
Additional Details : Gewicht : 0.01000kg
Beschreibung
PYHIN1 Polyclonal antibody specifically detects PYHIN1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PYHIN1 | |
Polyclonal | |
Unconjugated | |
PYHIN1 | |
IFIX; interferon-inducible protein X; PYHIN1; pyrin and HIN domain family member 1; pyrin and HIN domain family, member 1; pyrin and HIN domain-containing protein 1; RP11-520H16.1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
149628 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q6K0P9 | |
PYHIN1 | |
KLH conjugated synthetic peptide between 26-55 amino acids from the N-terminal region of human PYHIN1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |