Learn More
PYGM Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529132
Additional Details : Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A549, HeLa, Jurkat, Raji, mouse brain. Predicted reactivity: Mouse (94%), Rat (93%), Xenopus laevis (84%), Rabbit (95%), Sheep (96%), Rhesus Monkey (98%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PYGM Polyclonal antibody specifically detects PYGM in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PYGM | |
Polyclonal | |
Unconjugated | |
PYGM | |
AI115133; glycogen muscle; glycogen phosphorylase; glycogen phosphorylase, muscle associated; glycogen phosphorylase, muscle form; GPMM; muscle (McArdle syndrome); muscle glycogen phosphorylase; Muscpho; Myophosphorylase; PG; Phosphorylase glycogen; phosphorylase, glycogen, muscle; Phosphorylase, glycogen; muscle (McArdle syndrome); Pygm; unnamed protein product | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19309, 5837 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.26 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P11217, Q9WUB3 | |
PYGM | |
Recombinant fragment corresponding to a region within amino acids 511 and 753 of Human PYGM | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |