Learn More
PRK1 Mouse, Unlabeled, Clone: 49, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 610687
Additional Details : Gewicht : 0.01000kg
Beschreibung
Members of the Protein Kinase C (PKC) family of homologous serine/threonine protein kinases are involved in a number of processes such as cell growth, cell differentiation, and cytokine secretion. PKCs are activated by Ca2+, phospholipids, diacylglycerol, phorbol esters, and proteolysis. PRK1 (PKC-Related Kinase 1, also named PKN) was originally identified in human hippocampus as a novel protein kinase with sequence homology to PKC. PRK1 contains 942 amino acids with an apparent molecular weight of 120kDa. Although activated by limited proteolysis, PRK1 is not activated by Ca2+/diacylglycerol or phorbol esters. However, PRK1 is activated by phospholipids and arachidonic acid. PRK1 may regulate cytoskeletal changes since it binds to Rho-GTP and becomes phosphorylated in vivo, coincidentally with the formation of focal adhesions and stress fibers.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Spezifikation
PRK1 | |
Monoclonal | |
250μg/mL | |
PKN | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Western Blot | |
49 | |
Unconjugated | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
150 μg | |
Cell Biology | |
Canine, Human, Mouse, Rat | |
IgG1 |
For Research Use Only.