missing translation for 'onlineSavingsMsg'
Learn More

PRAK Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA528890

Additional Details : Gewicht : 0.01000kg

Artikelnummer. 13473007

  • 606.00 CHF / 100 Mikroliter
Verfügbar ab: 17-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Recommended positive controls: Raji. Predicted reactivity: Mouse (98%), Rat (97%), Zebrafish (93%), Pig (98%), Chicken (97%), Rhesus Monkey (99%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PRAK Polyclonal antibody specifically detects PRAK in Human samples. It is validated for Western Blot.
Spezifikation

Spezifikation

PRAK
Polyclonal
Unconjugated
MAPKAPK5
MAP kinase-activated protein kinase 5; MAPK-activated protein kinase 5; MAPKAP kinase 5; Mapkapk5; MAPKAP-K5; MAPKAPK-5; mitogen-activated protein kinase-activated protein kinase 5; MK5; MK-5; p38-regulated/activated protein kinase; PRAK
Rabbit
Antigen affinity chromatography
RUO
8550
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.64 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q8IW41
MAPKAPK5
Recombinant fragment corresponding to a region within amino acids 1 and 348 of Human PRAK
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote