Learn More
PP5 Mouse, Unlabeled, Clone: 3, BD
Mouse Monoclonal Antibody
547.01 CHF
Spezifikation
Antigen | PP5 |
---|---|
Klon | 3 |
Konzentration | 250μg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15825769
|
BD Biosciences
611021 |
150 μg |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
Beschreibung
Phosphorylation and dephosphorylation on serine and threonine residues is critical for signal transduction and the regulation of numerous cellular functions. Phosphorylation levels are modulated by protein kinases and phosphatases. The Ser/Thr phosphatase PP5 (also known as PPT) is a model protein of the fourth subfamily of the PPP-family of phosphatases which includes PP1, PP2A, and PP2B. PP5 is primarily located in the nucleus, demonstrates widespread tissue distribution, and is highly conserved among species. It consists of a C-terminal catalytic domain and an N-terminal TPR (tetratricopeptide repeat) domain which is involved in protein-protein interactions. The TPRs of PP5 mediate protein phosphatase activity. TPRs mediate the interaction of PP5 with various proteins including the hsp90-glucocorticoid receptor complex and the kinase domain of ANP guanylate cyclase receptor. PP5 is thought to be important in cell growth. For example, it functions upstream of p53 to regulate p21/WAF1/Cip1-mediated G1 growth arrest. Thus, PP5 is a nuclear protein phosphatase that is involved in signaling pathways of cell growth and arrest.
Immunofluorescence, Western Blotting
Spezifikation
PP5 | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified | |
PPT |
3 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Human, Mouse, Rat | |
Rat PP5 aa. 36-238 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.