Learn More
anti-PLCD4 Matched Antibody Pair, Abnova
Marke: Abnova H00084812-AP11.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-PLCD4 (100μg); Mouse monoclonal anti-PLCD4, IgG1κ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Beschreibung
Phosphatidylinositol-specific phospholipase C (PLC) plays an important role in receptor-mediated signal transduction by generating 2 second messenger molecules, inositol 1,4,5-triphosphate (IP3) and diacylglycerol, from phosphatidylinositol 4,5-bisphosphate (PIP2). PLC comprises a diverse family of enzymes that differ in structure and tissue distribution.
ELISA
Spezifikation
| PLCD4 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human PLCD4. | |
| MGC12837 | |
| Mouse, Rabbit | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-PLCD4 (100 μg) 2. Detection antibody: mouse monoclonal anti-PLCD4, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Unconjugated | |
| PLCD4 | |
| PLCD4 | |
| Affinity Purified | |
| RUO | |
| 84812 | |
| Human | |
| IgG1 |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.