Learn More
Phosphoserine Mouse, Unlabeled, Clone: 19/P-SER, BD
Mouse Monoclonal Antibody
520.00 CHF - 547.01 CHF
Spezifikation
Antigen | Phosphoserine |
---|---|
Klon | 19/P-SER |
Konzentration | 250μg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15816219
|
BD Biosciences
612546 |
50 μg |
520.00 CHF
1 Stück |
Verfügbar ab: 23-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15826219
|
BD Biosciences
612547 |
150 μg |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
Beschreibung
Protein phosphorylation of serine and threonine residues is critical for the control of protein activity involved in various cellular events. An assortment of Ser/Thr kinases and phosphatases regulate serine and threonine phosphorylation in cell signaling pathways, such as growth factor, cytokine, chemokine, and stress response. Detection of serine and threonine phosphorylation can generally be monitored by antibodies that detect phosphoserine and phosphothreonine. Our clone 19 antibody specifically recognizes phosphoserine modifications on peptides in ELISA, while our clone 22a detects both phosphoserine and phosphothreonine modifications on peptides in ELISA. These antibodies are reported to be useful for Western blot, flow cytometry, and microscopy detection of phosphoserine and phosphothreonine levels.
Immunofluorescence, Western Blotting
Spezifikation
Phosphoserine | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
19/P-SER | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Rat | |
Human Phosphoserine | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.