Learn More
Phospho-EGFR (Tyr992) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke:  Invitrogen PA517835
Additional Details : Gewicht : 0.01000kg
Artikelnummer. 11557080
  • 540.00 CHF / 100 Mikroliter
Verfügbar ab: 16-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
It is not recommended to aliquot this antibody.
Phospho-EGFR (Tyr992) Polyclonal antibody specifically detects Phospho-EGFR (Tyr992) in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Spezifikation
Spezifikation
Phospho-EGFR (Tyr992)
Polyclonal
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
P00533
EGFR
Synthetic phosphopeptide corresponding to residues surrounding pTyr992 of human EGF receptor
100 μL
Primary
Human, Rat
Antibody
IgG
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
EGFR
2.7.10.1; 9030024J15Rik; AI552599; avian erythroblastic leukemia viral (v-erbB) oncogene homolog; avian erythroblastic leukemia viral (v-erb-b) oncogene homolog; cell growth inhibiting protein 40; cell proliferation-inducing protein 61; EC 2.7.10.1; egf receptor; Egfr; EGFR-related peptide; epidermal growth factor receptor; epidermal growth factor receptor (erythroblastic leukemia viral (v-erb-b) oncogene homolog, avian); Epidermal growth factor receptor formerly avian erythroblastic leukemia viral (v-erbB) oncogene homolog (Erbb1); epidermal growth factor receptor, formerly avian erythroblastic leukemia viral (v-erbB) oncogene homolog (Erbb1); ERBB; ERBB1; ErbB-1; erb-b2 receptor tyrosine kinase 1; Errb1; Errp; HER1; kinase EGFR; mENA; NISBD2; Oncogene ERBB; PIG61; Proto-oncogene c-ErbB-1; receptor tyrosine-protein kinase erbB-1; Urogastrone; wa2; wa-2; Wa5; waved 2
Rabbit
Antigen affinity chromatography
RUO
1956, 24329
-20°C