missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-CD4 (Ser433) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538447
Additional Details : Gewicht : 0.00900kg
Beschreibung
The CD4 antigen is involved in the recognition of MHC class II molecules and is a co-receptor for HIV. CD4 is primarily expressed in a subset of T-lymphocytes, also referred to as T helper cells, but may also be expressed by other cells in the immune system, such as monocytes, macrophages, and dendritic cells. At the tissue level, CD4 expression may be detected in thymus, lymph nodes, tonsils, and spleen, and also in specific regions of the brain, gut, and other non-lymphoid tissues. CD4 functions to initiate or augment the early phase of T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase, Lck. It may also function as an important mediator of direct neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcripts have been identified in this gene [RefSeq, July 2017].Spezifikation
Phospho-CD4 (Ser433) | |
Polyclonal | |
Unconjugated | |
CD4 | |
CD4 antigen p55, CD4 receptor, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
920 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P01730 | |
CD4 | |
A synthetic phosphopeptide derived from human CD4 around the phosphorylation site of Ser433 (R-M-SP-Q-I) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |