missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-B-Raf (Ser445) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517535
Additional Details : Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody.
Phospho-B-Raf (Ser445) Polyclonal antibody specifically detects Phospho-B-Raf (Ser445) in Canine, Chicken, Human, Mouse, Non-human Primate, Rat samples. It is validated for Western Blot.Spezifikation
Phospho-B-Raf (Ser445) | |
Polyclonal | |
Unconjugated | |
BRAF | |
94 kDa B-raf protein; 9930012E13Rik; AA120551; AA387315; AA473386; Braf; B-raf; B-raf protein; B-raf protein isoform 1; B-raf protein isoform 2; B-Raf proto-oncogene serine/threonine-protein kinase; B-Raf proto-oncogene serine/threonine-protein kinase (p94); B-Raf proto-oncogene, serine/threonine kinase; Braf transforming gene; BRAF1; B-RAF1; Braf2; Braf-2; C230098H17; C87398; C-RMIL; D6Ertd631e; FLJ95109; MGC126806; murine sarcoma viral (v-raf) oncogene homolog B1; NS7; p94; Proto-oncogene B-Raf; proto-oncogene c-Rmil; RAFB1; RMIL; rmil serine/threonine-protein kinase; serine/threonine kinase; serine/threonine-protein kinase B-raf; Serine/threonine-protein kinase Rmil; v-raf murine sarcoma viral oncogene homolog B; v-raf murine sarcoma viral oncogene homolog B1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
109880, 114486, 396239, 475526, 673 | |
-20°C | |
Liquid |
Western Blot | |
4 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P15056, P28028, Q04982 | |
BRAF | |
Phospho synthetic peptide corresponding to residues surrounding serine 445 of human B-Raf | |
100 μL | |
Primary | |
Canine, Chicken, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |